Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

188239

Sigma-Aldrich

4-(4-Methoxyphenyl)-1-butanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4(CH2)4OH
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

refractive index

n20/D 1.526 (lit.)

bp

160-161 °C/8 mmHg (lit.)

mp

3-4 °C (lit.)

density

1.042 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CCCCO)cc1

InChI

1S/C11H16O2/c1-13-11-7-5-10(6-8-11)4-2-3-9-12/h5-8,12H,2-4,9H2,1H3

InChI key

ONIBHZIXCLTLNO-UHFFFAOYSA-N

General description

Decay of 4-(4-methoxyphenyl)-1-butanol radical cation in water via Cα-H deprotonation has been kinetically investigated by pulse radiolysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxygen Versus Carbon Acidity in the Side-Chain Fragmentation of 2-, 3-, and 4-Arylalkanol Radical Cations in Aqueous Solution: The Influence of the Distance between the OH Group and the Aromatic Ring1.
Baciocchi E, et al.
Journal of the American Chemical Society, 121(28), 6624-6629 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service