Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187895

Sigma-Aldrich

3-Bromobenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2OH
CAS Number:
Molecular Weight:
187.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.584 (lit.)

bp

165 °C/16 mmHg (lit.)

density

1.56 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

OCc1cccc(Br)c1

InChI

1S/C7H7BrO/c8-7-3-1-2-6(4-7)5-9/h1-4,9H,5H2

InChI key

FSWNRRSWFBXQCL-UHFFFAOYSA-N

Application

3-Bromobenzyl alcohol was employed in the preparation of tert-butyl 3-(3-bromophenyl)-2-cyanopropanoate and silyl ether.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocky J Barney et al.
Bioorganic & medicinal chemistry, 18(20), 7212-7220 (2010-09-14)
Geminal bisphosphonates display varied biological activity depending on the nature of the substituents on the central carbon atom. For example, the nitrogenous bisphosphonates zoledronate and risedronate inhibit the enzyme farnesyl diphosphate synthase while digeranyl bisphosphonate has been shown to inhibit
Iridium catalysed alkylation of 4-hydroxy coumarin, 4-hydroxy-2-quinolones and quinolin-4 (1H)-one with alcohols under solvent free thermal conditions.
Grigg R, et al.
Tetrahedron, 65(36), 7468-7473 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service