Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187291

Sigma-Aldrich

2-Methyl-3-nitrobenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

69-73 °C (lit.)

functional group

hydroxyl
nitro

SMILES string

Cc1c(CO)cccc1[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-7(5-10)3-2-4-8(6)9(11)12/h2-4,10H,5H2,1H3

InChI key

QJANIQCEDPJNLO-UHFFFAOYSA-N

Application

2-Methyl-3-nitrobenzyl alcohol was used to prepare 2-methyl-3-nitrobenzyl cyanide. It was also used in the synthesis of 20-membered macrocyclic thiodepsipeptide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation and Claisen condensation products of 3-nitro-o-xylene.
Askam V and Deeks RHL.
J. Chem. Soc. Sect. C, 14, 1935-1936 (1969)
Marc C Kimber et al.
Chemical communications (Cambridge, England), (5)(5), 591-593 (2008-01-23)
A 20-membered macrocyclic thiodepsipeptide has been synthesized as a model for the southern hemisphere of nosiheptide, the key steps being assembly of an acyclic precursor by amide coupling of indole and thiazole fragments followed by formation of the thiolactone in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service