Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187127

Sigma-Aldrich

1,9-Diaminononane

98%

Synonym(s):

1,9-Nonanediamine, Nonamethylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$303.00

$303.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$303.00

About This Item

Linear Formula:
NH2(CH2)9NH2
CAS Number:
Molecular Weight:
158.28
Beilstein:
1737533
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$303.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

258-259 °C/756 mmHg (lit.)

mp

35-37 °C (lit.)

SMILES string

NCCCCCCCCCN

InChI

1S/C9H22N2/c10-8-6-4-2-1-3-5-7-9-11/h1-11H2

InChI key

SXJVFQLYZSNZBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Soo Jo et al.
Polymers, 11(5) (2019-05-15)
A set of poly(pyridinium) salts containing various diamine moieties, as molecular connectors, and poly(ethyleneglycol)-4-nonylphenyl-3-sulfopropyl ether, thereafter referred to as "Macroion", as the hydrophilic counterion, were prepared by metathesis reaction from the respective precursory tosylated poly(pyridinium)s in methanol. The structure of
Fátima M F Vergara et al.
Bioorganic & medicinal chemistry letters, 19(17), 4937-4938 (2009-08-04)
A series of 11 alpha,omega-diaminoalkanes, (H(2)N(CH(2))(n)NH(2), n=2-12) have been evaluated for their in vitro antibacterial activity against Mycobacterium tuberculosis H37Rv. Compounds, (H(2)N(CH(2))(n)NH(2), n=9-12), exhibited a very good activities in the range 2.50-3.12 microg/mL, which can be compared with that of
Huai-Hsin Su et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 978-979, 131-137 (2014-12-30)
Spermine and spermidine are multiple-nitrogen compounds found in many foods. Both compounds are essential for cell growth and human health. This study established a simple and fast method of detecting spermine and spermidine in food samples by matrix-assisted laser desorption/ionization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service