Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

187119

Sigma-Aldrich

3,4,5-Trichloroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2NH2
CAS Number:
Molecular Weight:
196.46
Beilstein:
638880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

97-99 °C (lit.)

SMILES string

Nc1cc(Cl)c(Cl)c(Cl)c1

InChI

1S/C6H4Cl3N/c7-4-1-3(10)2-5(8)6(4)9/h1-2H,10H2

InChI key

XOGYQVITULCUGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4,5-Trichloroaniline forms tri- and tetrachloroaniline-Pt(II) complexes having good antileukemic activity.

Application

3,4,5-Trichloroaniline was used in the determination of substituted aromatic amines in sediment or water samples by capillary column gas chromatography.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Craciunescu et al.
Chemico-biological interactions, 53(1-2), 45-56 (1985-02-01)
The paper reports the synthesis, the chemical characterization and the IR data of new Pt(II) and Pt(IV) complexes, as well as their cytostatic activities on KB cells and antitumour properties against three tumour systems (P388 and L1210 leukemias and advanced
The determination of substituted aromatic amines in water and sediment samples.
Scholz B and Palauschek N.
Fresenius' Zeitschrift fur Analytische Chemie, 331(3-4), 282-289 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service