Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

186902

Sigma-Aldrich

2,6-Dimethoxybenzonitrile

97%

Synonym(s):

2-Cyanoresorcinol dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Assay

97%

SMILES string

COc1cccc(OC)c1C#N

InChI

1S/C9H9NO2/c1-11-8-4-3-5-9(12-2)7(8)6-10/h3-5H,1-2H3

InChI key

XHAHKSSLDJIEDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service