Skip to Content
MilliporeSigma
All Photos(2)

Documents

186686

Sigma-Aldrich

7-(Trifluoromethyl)quinoline-4-thiol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F3NS
CAS Number:
Molecular Weight:
229.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

chunks

mp

222-225 °C (lit.)

SMILES string

FC(F)(F)c1ccc2c(S)ccnc2c1

InChI

1S/C10H6F3NS/c11-10(12,13)6-1-2-7-8(5-6)14-4-3-9(7)15/h1-5H,(H,14,15)

InChI key

DBWDEWRMEKXOFI-UHFFFAOYSA-N

Application

7-(Trifluoromethyl)quinoline-4-thiol was used to investigate the mechanism of generation of lipid-poor particles from high-density lipoprotein induced by cholesteryl ester transfer protein.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyrille Maugeais et al.
Biochimica et biophysica acta, 1831(11), 1644-1650 (2013-07-23)
Cholesteryl ester transfer protein (CETP), a key regulator of high-density lipoprotein (HDL) metabolism, induces HDL remodeling by transferring lipids between apolipoprotein B-containing lipoproteins and HDL, and/or by promoting lipid transfer between HDL subparticles. In this study, we investigated the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service