Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

186597

Sigma-Aldrich

Dimethyl 3-methylglutaconate, mixture of cis and trans

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCH2C(CH3)=CHCO2CH3
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.456 (lit.)

bp

109-110 °C/12 mmHg (lit.)

density

1.095 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C\C(C)=C\C(=O)OC

InChI

1S/C8H12O4/c1-6(4-7(9)11-2)5-8(10)12-3/h4H,5H2,1-3H3/b6-4+

InChI key

GBQBEQOEGMZCOH-GQCTYLIASA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service