Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

185477

Sigma-Aldrich

Isopropyl acetate

99%

Synonym(s):

Acetic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740761
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

47 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

894 °F

expl. lim.

1.8 %, 37 °F
8 %

refractive index

n20/D 1.377 (lit.)

bp

85-91 °C (lit.)

mp

−73 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)=O

InChI

1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3

InChI key

JMMWKPVZQRWMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thu Quach et al.
American journal of public health, 101 Suppl 1, S271-S276 (2011-05-10)
We engaged Vietnamese nail salon workers in a community-based participatory research (CBPR) study to measure personal and area concentrations of solvents in their workplace. We measured average work-shift concentrations of toluene, ethyl acetate, and isopropyl acetate among 80 workers from
R Akimoto et al.
The Journal of surgical research, 53(6), 572-577 (1992-12-01)
Methyl tert-butyl ether (MTBE) is an organic solvent that has been used to dissolve gallstones via a percutaneous transhepatic catheter into the gallbladder. To test whether MTBE might cause serious tissue injury if accidentally infused outside the gallbladder, the effect
Vapor-liquid equilibria of carbon dioxide with isopropyl acetate, diethyl carbonate and ethyl butyrate at elevated pressures.
Cheng CH and Chen YP.
Fluid Phase Equilibria, 234(1), 77-83 (2005)
General study of catalytic oxidation of various VOCs over La0.8Sr0.2MnO3+x perovskite catalyst-influence of mixture.
Blasin-Aube V, et al.
Applied Catalysis. B, Environmental, 43(2), 175-186 (2003)
A novel route to produce thymol by vapor phase reaction of m-cresol with isopropyl acetate over Al-MCM-41 molecular sieves.
Shanmugapriya K, et al.
J. Catal., 224(2), 347-357 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service