Skip to Content
MilliporeSigma
All Photos(1)

Documents

185272

Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate

97%

Synonym(s):

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, Dimethyl succinylsuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O6
CAS Number:
Molecular Weight:
228.20
Beilstein:
3345562
EC Number:
MDL number:
PubChem Substance ID:

Assay

97%

SMILES string

COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

InChI

1S/C10H12O6/c1-15-9(13)5-3-8(12)6(4-7(5)11)10(14)16-2/h5-6H,3-4H2,1-2H3

InChI key

MHKKFFHWMKEBDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service