Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184594

Sigma-Aldrich

Poly(methyl vinyl ether-alt-maleic anhydride)

average Mw ~1,980,000 by LS, average Mn ~960,000, powder

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H2O3 · C3H6O)n
CAS Number:
MDL number:
UNSPSC Code:
12162002

form

powder

mol wt

average Mn ~960,000
average Mw ~1,980,000 by LS

impurities

<2% benzene

solubility

H2O: soluble (upon hydrolysis)
organic solvents: soluble

InChI

1S/C4H2O3.C3H6O/c5-3-1-2-4(6)7-3;1-3-4-2/h1-2H;3H,1H2,2H3

InChI key

UPBDXRPQPOWRKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

Alternating copolymer.

comparable product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service