Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

183997

Sigma-Aldrich

Trihexylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]3N
CAS Number:
Molecular Weight:
269.51
Beilstein:
1755828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.3 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

150-159 °C/12 mmHg (lit.)
263-265 °C (lit.)

density

0.794 g/mL at 25 °C (lit.)

SMILES string

CCCCCCN(CCCCCC)CCCCCC

InChI

1S/C18H39N/c1-4-7-10-13-16-19(17-14-11-8-5-2)18-15-12-9-6-3/h4-18H2,1-3H3

InChI key

DIAIBWNEUYXDNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trihexylamine was used as extractant during isolation and purification of succinic acid from an Escherichia coli fermentation broth. It was employed as internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. It was used in the synthesis of small diameter zinc oxide nanorods.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T Keller et al.
Forensic science international, 94(1-2), 55-63 (1998-07-22)
Since its inception in the early 1970s under the name plasma chromatography, ion mobility spectrometry (IMS) has undergone great changes. It is now utilized more and more in forensic science laboratories where it is used to detect explosives and environmental
Tanja Kurzrock et al.
Biotechnology progress, 27(6), 1623-1628 (2012-01-13)
Biotechnologically produced succinic acid has the potential to displace maleic acid and its uses. Therefore, it is of high interest for the chemical, pharmaceutical, and food industry.In addition to optimized production strains and fermentation processes, an efficient separation of succinic
Louwanda Lakiss et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(16), 1873-1881 (2020-03-17)
We outline two methodologies to selectively characterize the Brønsted acidity of the external surface of FAU-type zeolites by IR and NMR spectroscopy of adsorbed basic probe molecules. The challenge and goal are to develop reliable and quantitative IR and NMR
Diameter control and photoluminescence of ZnO nanorods from trialkylamines.
Andelman T, et al.
Journal of Nanomaterials, 2007 (2007)
Erica L Bakota et al.
Rapid communications in mass spectrometry : RCM, 34(5), e8608-e8608 (2019-11-11)
While liquid chromatography/high-resolution mass spectrometry (LC/HRMS) is a versatile analytical technique, it is also sensitive to trace impurities. These impurities may come from a variety of sources, including reagents, solvents, and the sample matrix itself. Impurities in reagents may become

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service