Skip to Content
MilliporeSigma
All Photos(1)

Documents

183776

Sigma-Aldrich

N-Methyldioctylamine

98%

Synonym(s):

N,N-Dioctylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]2NCH3
CAS Number:
Molecular Weight:
255.48
Beilstein:
1750823
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.4424 (lit.)

bp

162-165 °C/15 mmHg (lit.)

mp

−30.1 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN(C)CCCCCCCC

InChI

1S/C17H37N/c1-4-6-8-10-12-14-16-18(3)17-15-13-11-9-7-5-2/h4-17H2,1-3H3

InChI key

YJLYANLCNIKXMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service