Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

183458

Sigma-Aldrich

1-Naphthaleneethanol

95%

Synonym(s):

2-(1-Naphthyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CH2OH
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

bp

186 °C/17 mmHg (lit.)

mp

62-65 °C (lit.)

functional group

hydroxyl

SMILES string

OCCc1cccc2ccccc12

InChI

1S/C12H12O/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7,13H,8-9H2

InChI key

RXWNCMHRJCOWDK-UHFFFAOYSA-N

Gene Information

human ... BAD(572)

Looking for similar products? Visit Product Comparison Guide

Packaging

Total impurities <5% 2-isomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianlin Wang et al.
Journal of chromatography. A, 1013(1-2), 19-27 (2003-11-08)
A modified field amplification sample injection method was proposed and evaluated by using positively mono charged cyclodextrin (CD) as carrier and 1-adamantanecarboxylate as displacer for on-capillary preconcentration of neutral compounds and improvement of the concentration limit of detection in micellar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service