Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

18289

Sigma-Aldrich

1-Dodecyl-3-methylimidazolium iodide

≥95.0%

Synonym(s):

1-Lauryl-3-methylimidazolium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31IN2
CAS Number:
Molecular Weight:
378.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

impurities

≤0.5% water

storage temp.

2-8°C

SMILES string

[I-].CCCCCCCCCCCCn1cc[n+](C)c1

InChI

1S/C16H31N2.HI/c1-3-4-5-6-7-8-9-10-11-12-13-18-15-14-17(2)16-18;/h14-16H,3-13H2,1-2H3;1H/q+1;/p-1

InChI key

RQVJLVQTNZVVRZ-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kiran Kumar Reddy et al.
Frontiers in microbiology, 11, 730-730 (2020-05-07)
Candida albicans is an opportunistic pathogen causes fungal infections that range from common skin infections to persistent infections through biofilm formation on tissues, implants and life threatening systemic infections. New antifungal agents or therapeutic methods are desired due to high

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service