Skip to Content
MilliporeSigma
All Photos(1)

Documents

182036

Sigma-Aldrich

Poly(ethylene succinate)

average Mw 10,000

Synonym(s):

Ethylene glycol succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OCH2CH2O2CCH2CH2CO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

chunks
crystals
granular

mol wt

average Mw 10,000

transition temp

Tg −1 °C
Tm 108 °C

SMILES string

OCCO.OC(=O)CCC(O)=O

InChI

1S/C4H6O4.C2H6O2/c5-3(6)1-2-4(7)8;3-1-2-4/h1-2H2,(H,5,6)(H,7,8);3-4H,1-2H2

InChI key

VJVOPINBJQWMNY-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin Charnock
Microorganisms, 9(1) (2021-01-07)
Plastic pollution has become one of the most critical environmental issues, as rapidly increasing production, compounded by persistence of plastic wastes in the environment, are outpacing efforts to keep ecosystems plastic-free. A switch to plastics more amenable to microbial attack

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service