Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181544

Sigma-Aldrich

Poly(isobutyl methacrylate)

average Mw ~70,000, crystalline

Synonym(s):

Isobutyl methacrylate homopolymer, iso -Butyl methacrylate polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H14O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

crystalline

mol wt

average Mw ~70,000

impurities

<1.5% isobutyl methacrylate
<2% toluene

refractive index

n20/D 1.477

transition temp

Tg 55 °C

density

1.09 g/mL at 25 °C

SMILES string

CC(C)COC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PiBMA can be prepared by free radical polymerization.

Application

Poly(isobutyl methacrylate) (PiBMA) was used as a solvent in thin layer chromatography. Glycidyl methacrylate grafted poly(isobutyl methacrylate) (PiBMA) microbeads can be used as super adsorbent beads for adsorption of heavy metal ions such as uranyl ions from aqueous solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of poly (isobutyl methacrylate) microbeads with grafted amidoxime groups.
Caykara T, et al.
Radiation Physics and Chemistry, 76(10), 1569-1576 (2007)
Determination of glycolipid?protein interaction specificity.
Lopez P H and Schnaar R L
Methods in Enzymology, 417, 205-220 (2006)
Licinia Santos et al.
Glycobiology, 30(11), 881-894 (2020-04-14)
The FORS histo-blood group system is the most recently discovered carbohydrate-based human blood group system. FORS is a rare blood group system, and most individuals have naturally occurring anti-FORS1 antibodies in plasma. Screening for anti-FORS1 antibodies is often done by
Janaina Habib Jorge et al.
Quintessence international (Berlin, Germany : 1985), 40(10), e93-100 (2010-02-18)
To evaluate the effect of microwave postpolymerization heat treatment and water storage time on the cytotoxicity of denture base and reline acrylic resins. Sample disks of 6 acrylic resins were fabricated under aseptic conditions (Kooliner, GC; Tokuyama Rebase II, Tokuyama
A G Zuccari et al.
Bio-medical materials and engineering, 7(5), 327-343 (1997-01-01)
Acrylic resins are commonly used in many dental applications; especially in the fabrication of provisional fixed partial dentures. Among noticed technical drawbacks associated with this material are unsatisfactory mechanical properties. Moreover, if acrylic resins are exposed to moist environment, water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service