Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181277

Sigma-Aldrich

Poly(acrylamide-co-acrylic acid)

average Mw 5,000,000, powder

Synonym(s):

Acrylamide-acrylic acid copolymer, Acrylamide-acrylic acid resin, Poly(acrylamide-acrylic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

powder

mol wt

average Mw 5,000,000

impurities

10-15 wt. % water
5-10 wt. % sodium sulfate

viscosity

2-3 cP, 0.1 % in H2O

transition temp

Tg 165 °C

solubility

water and some protic solvents: soluble

density

0.75 g/mL at 25 °C (lit.)

SMILES string

NC(=O)C=C.OC(=O)C=C

InChI

1S/C3H5NO.C3H4O2/c2*1-2-3(4)5/h2H,1H2,(H2,4,5);2H,1H2,(H,4,5)

InChI key

RNIHAPSVIGPAFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thickening agent, binder, sizing agent, flocculating agent, crosslinker, filtering aid, lubricant, drag reduction and polymer recovery agent.

Features and Benefits

Changes in pH from 3 to 10 have little effect on viscosity. High tolerance for electrolytes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service