Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18072

Sigma-Aldrich

(S)-(−)-1-(4-Bromophenyl)ethylamine

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-4-Bromo-α-methylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
Beilstein:
5729988
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D −20.5±1°, c = 3% in methanol

refractive index

n20/D 1.567

density

1.390 g/mL at 20 °C (lit.)

SMILES string

C[C@H](N)c1ccc(Br)cc1

InChI

1S/C8H10BrN/c1-6(10)7-2-4-8(9)5-3-7/h2-6H,10H2,1H3/t6-/m0/s1

InChI key

SOZMSEPDYJGBEK-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-1-(4-Bromophenyl)ethylamine may be used in the synthesis of (S)-1-(1-(4-bromophenyl) ethyl)-2-cyano-3-(quinoline-5-yl) guanidine, which is an intermediate to prepare a potent and selective antagonist and radioligand for rat P2X7 receptors.[1]

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[3H] A-804598 ([3H] 2-cyano-1-[(1S)-1-phenylethyl]-3-quinolin-5-ylguanidine) is a novel, potent, and selective antagonist radioligand for P2X7 receptors.
Donnelly-Roberts DL, et al.
Neuropharmacology, 56(1), 223-229 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service