Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18055

Sigma-Aldrich

4-Bromo-L-phenylalanine

≥98.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BrNO2
CAS Number:
Molecular Weight:
244.09
Beilstein:
2212159
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

solid

optical activity

[α]20/D −22.0±2°, c = 0.5% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

~265 °C (dec.)

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1ccc(Br)cc1)C(O)=O

InChI

1S/C9H10BrNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m0/s1

InChI key

PEMUHKUIQHFMTH-QMMMGPOBSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dose fractionation in neutron capture therapy for malignant melanoma.
B J Allen et al.
Basic life sciences, 50, 63-67 (1989-01-01)
James M Turner et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(17), 6483-6488 (2006-04-19)
Recently, tRNA aminoacyl-tRNA synthetase pairs have been evolved that allow one to genetically encode a large array of unnatural amino acids in both prokaryotic and eukaryotic organisms. We have determined the crystal structures of two substrate-bound Methanococcus jannaschii tyrosyl aminoacyl-tRNA
M Ibba et al.
FEBS letters, 364(3), 272-275 (1995-05-15)
It has previously been demonstrated that the unnatural amino acid p-Cl-phenylalanine can be attached to tRNA(Phe) by a modified phenylalanyl-tRNA synthetase with relaxed amino acid substrate specificity. We show that this modification to the translational machinery of Escherichia coli is
N Sharma et al.
FEBS letters, 467(1), 37-40 (2000-02-09)
Artificial proteins can be engineered to exhibit interesting solid state, liquid crystal or interfacial properties and may ultimately serve as important alternatives to conventional polymeric materials. The utility of protein-based materials is limited, however, by the availability of just the
Neutron capture therapy for melanoma.
J A Coderre et al.
Basic life sciences, 50, 219-232 (1989-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service