Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

180033

Sigma-Aldrich

4-Methoxy-N-methylaniline

98%

Synonym(s):

N-Methyl-p-anisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NHCH3
CAS Number:
Molecular Weight:
137.18
Beilstein:
774640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

135-136 °C/19 mmHg (lit.)

mp

33-36 °C (lit.)

functional group

amine

SMILES string

CNc1ccc(OC)cc1

InChI

1S/C8H11NO/c1-9-7-3-5-8(10-2)6-4-7/h3-6,9H,1-2H3

InChI key

JFXDIXYFXDOZIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The allylation of N-methyl-p-anisidine by quaternary allylammonium cation was studied[1].

Application

4-Methoxy-N-methylaniline (N-Methyl-p-anisidine) was used to study the additive effects of amines[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Cappelli et al.
Bioorganic & medicinal chemistry, 16(6), 3428-3437 (2008-02-26)
The exploration of the structure-affinity relationships concerning a new class of peripheral benzodiazepine receptor (PBR) ligands related to alpidem has been pursued in order to evaluate the consistency of the structure-affinity relationships among different classes (and subclasses) of PBR ligands.
Takuto Nagano et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(37), 11578-11592 (2012-08-24)
The additive effects of amines were realized in the asymmetric hydrogenation of 2-phenylquinoxaline, and its derivatives, catalyzed by chiral cationic dinuclear triply halide-bridged iridium complexes [{Ir(H)[diphosphine]}(2)(μ-X)(3)]X (diphosphine = (S)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl [(S)-BINAP], (S)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole [(S)-SEGPHOS], (S)-5,5'-bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole [(S)-DIFLUORPHOS]; X = Cl, Br, I) to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service