Skip to Content
MilliporeSigma
All Photos(1)

Documents

178039

Sigma-Aldrich

2-Chloro-6-fluorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CO2H
CAS Number:
Molecular Weight:
174.56
Beilstein:
973857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

155-157 °C (lit.)

SMILES string

OC(=O)c1c(F)cccc1Cl

InChI

1S/C7H4ClFO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H,10,11)

InChI key

XNTIGDVFBDJLTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-6-fluorobenzoic acid is a two-fold halogenated derivative of benzoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Betz et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 6), o1329-o1329 (2011-07-15)
The title compound, C(7)H(4)ClFO(2), is a twofold halogenated derivative of benzoic acid. The C-C-C angles within the aromatic moiety cover a range 116.11 (14)-123.96 (15)°, with the maximum and the minimum value next to each other. In the crystal, O-H⋯O hydrogen bonds
Gulce Ogruc Ildiz et al.
Molecules (Basel, Switzerland), 25(21) (2020-10-30)
This article presents a detailed comprehensive investigation of the ortho fluoro- and chloro- substituted benzoic acids both, as isolated molecules and in the crystalline phase. Quantum chemical calculations performed within the density functional theory (DFT) formalism are used to investigate
David Plouffe et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(26), 9059-9064 (2008-06-27)
The growing resistance to current first-line antimalarial drugs represents a major health challenge. To facilitate the discovery of new antimalarials, we have implemented an efficient and robust high-throughput cell-based screen (1,536-well format) based on proliferation of Plasmodium falciparum (Pf) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service