Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177679

Sigma-Aldrich

7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

98%

Synonym(s):

5-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 6-Methyl-4-hydroxy-1,3,3a,7-tetraazaindene, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, NSC 32071, NSC 511493

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O
CAS Number:
Molecular Weight:
150.14
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

280-283 °C (lit.)

SMILES string

Cc1cc(O)n2ncnc2n1

InChI

1S/C6H6N4O/c1-4-2-5(11)10-6(9-4)7-3-8-10/h2-3,11H,1H3

InChI key

ZUIVNYGZFPOXFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for the synthesis of:
  • Ruthenium(II)-Hmtpo complexes
  • Dihydroorotate dehydrogenase inhibitors with antimalarial activity

Reactant for the Vilsmeier reaction of conjugated carbocycles and heterocycles

Investigations of the pharmacological activity caused by binding to HIV TAR RNA

Additive to study the space charge layer in silver bromide microcrystals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service