Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177407

Sigma-Aldrich

cis-3-Chloroacrylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CHCO2H
CAS Number:
Molecular Weight:
106.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

61-62 °C (lit.)

SMILES string

OC(=O)\C=C/Cl

InChI

1S/C3H3ClO2/c4-2-1-3(5)6/h1-2H,(H,5,6)/b2-1-

InChI key

MHMUCYJKZUZMNJ-UPHRSURJSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frederick Stull et al.
Nature structural & molecular biology, 23(1), 53-58 (2015-12-01)
Chaperones assist in the folding of many proteins in the cell. Although the most well-studied chaperones use cycles of ATP binding and hydrolysis to assist in protein folding, a number of chaperones have been identified that promote folding in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service