Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

175250

Sigma-Aldrich

6-Methoxy-1-indanone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
1238602
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

solid

mp

105-109 °C (lit.)

functional group

ketone

SMILES string

COc1ccc2CCC(=O)c2c1

InChI

1S/C10H10O2/c1-12-8-4-2-7-3-5-10(11)9(7)6-8/h2,4,6H,3,5H2,1H3

InChI key

UJGDLLGKMWVCPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Methoxy-1-indanone was used in the synthesis of 5-methoxyninhydrin (2,2-dihydroxy-5-methoxy-1,3-indanedione).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Routes to Ninhydrins. Preparation of Ninhydrin, 5-Methoxyninhydrin, and 5-(Methylthio) ninhydrin.
Synthetic Communications, 21(21), 2231-2256 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service