Skip to Content
MilliporeSigma
All Photos(1)

Documents

175129

Sigma-Aldrich

Antimony(V) fluoride

99%

Synonym(s):

Antimony pentafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbF5
CAS Number:
Molecular Weight:
216.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

2.2 (vs air)

vapor pressure

10 mmHg ( 25 °C)

Assay

99%

form

liquid

reaction suitability

reagent type: catalyst
core: antimony

bp

148-150 °C (lit.)

mp

7 °C (lit.)

solubility

liquid sulfur dioxide: slightly soluble(lit.)
potassium fluoride: slightly soluble(lit.)

density

2.993 g/mL at 25 °C (lit.)

SMILES string

F[Sb](F)(F)(F)F

InChI

1S/5FH.Sb/h5*1H;/q;;;;;+5/p-5

InChI key

VBVBHWZYQGJZLR-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

HF/SbF5 is an excellent medium for the preparation of alkylidene oxonium salts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rolf Minkwitz et al.
Inorganic chemistry, 38(5), 844-847 (2001-10-24)
We have studied the preparation and isolation of the halogenated alkylidene oxonium salts CH(3)OCHX(+)MF(6)(-) (X = Cl, F; M = As, Sb). Addition of dichloromethylmethylether at -78 degrees C to the superacidic media HF/MF(5) resulted in the formation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service