Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

173452

Sigma-Aldrich

4-(Phenylazo)benzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4COCl
CAS Number:
Molecular Weight:
244.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

94-97 °C (lit.)

SMILES string

ClC(=O)c1ccc(cc1)\N=N\c2ccccc2

InChI

1S/C13H9ClN2O/c14-13(17)10-6-8-12(9-7-10)16-15-11-4-2-1-3-5-11/h1-9H/b16-15+

InChI key

RYMHZBAYPLCCAC-FOCLMDBBSA-N

Application

4-(Phenylazo)benzoyl chloride was used in the synthesis of polyacetylene derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antje Modrow et al.
Dalton transactions (Cambridge, England : 2003), 41(28), 8690-8696 (2012-06-14)
For the first time an azo functionality was covalently introduced into a MOF by post-synthetic modification. The reaction of Cr-MIL-101-NH(2) with p-phenylazobenzoylchloride (1) and 4-(phenylazo)phenylisocyanate (2) as the reactants led to the compounds Cr-MIL-101_amide and Cr-MIL-101_urea, with the azo groups
Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 11(8), 7386-7389 (2011-11-23)
A new polyacetylene derivative was prepared by the activated polymerization of 2-ethynylpyridine by using 4-(phenylazo)benzoyl chloride without any additional initiator or catalyst in high yield. The chemical structure of poly[2-ethynyl-N-(4-(phenylazo)benzoyl) pyridinium chloride [PEPABPC] was characterized by such instrumental methods as
Santanu Mondal et al.
ACS chemical biology, 13(4), 1057-1065 (2018-03-09)
Protein arginine deiminases (PADs) play an important role in the pathogenesis of various diseases, including rheumatoid arthritis, multiple sclerosis, lupus, ulcerative colitis, and breast cancer. Therefore, the development of PAD inhibitors has drawn significant research interest in recent years. Herein
Weiyi Zhao et al.
Journal of controlled release : official journal of the Controlled Release Society, 323, 24-35 (2020-04-14)
Supramolecular hydrogels confer control over structural properties in a reversible, dynamic, and biomimetic fashion. The design of supramolecular hydrogels with an improved structural and functional recapitulation of damaged organs is important for clinical applications. For wound healing management, in particular
Beibei Hou et al.
Materials science & engineering. C, Materials for biological applications, 74, 94-102 (2017-03-04)
How to encapsulate and transport the payload of multiple therapeutic compounds avoiding premature leakage, and simultaneously co-release them rapidly at specific lesions still remains the major concern in clinic. Herein, we designed the UCN@mSiO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service