Skip to Content
MilliporeSigma
All Photos(1)

Documents

16819

Sigma-Aldrich

1-(3-Cyanopropyl)pyridinium bis(trifluoromethylsulfonyl)imide

≥98.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11F6N3O4S2
CAS Number:
Molecular Weight:
427.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.5% (T)

impurities

≤0.1% water
≤10 mg/kg halogenes

refractive index

n20/D 1.458±0.001

SMILES string

N#CCCC[n+]1ccccc1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

General description

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service