Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

167630

Sigma-Aldrich

4-Chloro-1,3-dioxolan-2-one

90%

Synonym(s):

Chloroethylene carbonate, Chloroethyleneglycol carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3ClO3
CAS Number:
Molecular Weight:
122.51
Beilstein:
109433
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

refractive index

n20/D 1.454 (lit.)

bp

121-123 °C/18 mmHg (lit.)

density

1.504 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC1COC(=O)O1

InChI

1S/C3H3ClO3/c4-2-1-6-3(5)7-2/h2H,1H2

InChI key

OYOKPDLAMOMTEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-1,3-dioxolan-2-one reacts with potassium fluoride under severe conditions to yield 4-fluoro-1,3-dioxolan-2-one.

Other Notes

Remainder ethylene carbonate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of direct fluorination technology for application to materials for lithium battery.
Kobayashi M, et al.
Journal of Fluorine Chemistry, 120(2), 105-110 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service