Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164623

Sigma-Aldrich

Tropylium tetrafluoroborate

97%

Synonym(s):

Cycloheptatrienyl tetrafluoroborate, Tropylium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BF4
CAS Number:
Molecular Weight:
177.94
Beilstein:
3574512
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

~240 °C (dec.) (lit.)

SMILES string

F[B-](F)(F)F.c1ccc[cH+]cc1

InChI

1S/C7H7.BF4/c1-2-4-6-7-5-3-1;2-1(3,4)5/h1-7H;/q+1;-1

InChI key

SQVQHTIKOZVGLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Retro-Buchner reactions for synthesis of arylcyclopropanes
  • Desymmetrization for access to homocalystegine analogs or aminocycloheptitols
  • α-Cyanation of imines
  • Stereoselective α-alkylation of aldehydes with stable carbocations
  • Oxidative C-H activation of aliphatic aldehydes
  • Electrochemical Redox transformations

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Banks et al.
Biochemical and biophysical research communications, 140(2), 609-615 (1986-10-30)
The stabilized carbonium ion salt, tropylium tetrafluoroborate, was oxidized to tropone (cycloheptatrienone) by rabbit liver aldehyde oxidase but not by the closely related molybdenum hydroxylase, xanthine oxidase. The tropylium cation is an aromatic hydrocarbon which lacks the aldehyde, imine, or

Articles

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service