Skip to Content
MilliporeSigma
All Photos(1)

Documents

164496

Sigma-Aldrich

2-(2-Ethoxyethoxy)ethanol-OD

97 atom % D

Synonym(s):

2-(2-Ethoxyethoxy)ethan(ol-d)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2OD
CAS Number:
Molecular Weight:
135.18
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

97 atom % D

form

liquid

refractive index

n20/D 1.426 (lit.)

density

1.006 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OCCOCCOCC

InChI

1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3/i7D

InChI key

XXJWXESWEXIICW-WHRKIXHSSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service