Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164313

Sigma-Aldrich

3-Methoxytyramine hydrochloride

96%

Synonym(s):

3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
3631283
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

96%

mp

213-215 °C (lit.)

SMILES string

Cl[H].COc1cc(CCN)ccc1O

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H

InChI key

AWRIOTVUTPLWLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A major metabolite of L-DOPA.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anouk N A Van Der Horst-Schrivers et al.
Anticancer research, 30(12), 5153-5158 (2010-12-29)
This study aimed to determine the prevalence of excess dopamine in relation to clinical symptoms and nuclear imaging in head and neck paraganglioma (PGL) patients. Thirty-six consecutive patients with head and neck PGLs, evaluated between 1993 and 2009, were included.
Moon Chul Jung et al.
Analytical chemistry, 78(6), 1761-1768 (2006-03-16)
The photoluminescence-following electron-transfer (PFET) technique, developed in our laboratory, is a sensitive chromatographic detection method for oxidizable analytes. Because the oxidations are homogeneous, the technique avoids the problem of electrode fouling. A liquid-phase oxidant reacts with the electrochemically active analytes
Wilhelmina H A de Jong et al.
The Journal of clinical endocrinology and metabolism, 94(8), 2841-2849 (2009-07-02)
Measurements of the 3-O-methylated metabolites of catecholamines [metanephrines (MNs)] in plasma or urine are recommended for diagnosis of pheochromocytoma. It is unclear whether these tests are susceptible to dietary influences. The aim of the study was to determine the short-term
Marc Morissette et al.
Journal of neuroscience research, 87(7), 1634-1644 (2008-12-31)
A higher prevalence and incidence of Parkinson's disease is observed in men, and beneficial motor effects of estrogens are observed in parkinsonian women. In rodents, an effect of estradiol on dopamine systems is documented, whereas much less is known in
Amal Alachkar et al.
Neuroscience research, 67(3), 245-249 (2010-03-23)
The ability of l-3,4-dihydroxyphenylalanine (l-DOPA), l-DOPA-methyl ester and their major metabolites, dopamine, dihydroxyphenylacetic acid (DOPAC), homovanillic (HVA), 3-O-methyldopa and 3-methoxytyramine (3-MT) to bind to alpha(2) adrenergic and D1 and D2 dopamine receptors was assessed by radioligand binding to cloned human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service