Skip to Content
MilliporeSigma
All Photos(1)

Documents

163252

Sigma-Aldrich

3,4-Dimethoxytoluene

96%

Synonym(s):

4-Methylveratrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OCH3)2
CAS Number:
Molecular Weight:
152.19
Beilstein:
2045328
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.528 (lit.)

bp

133-135 °C/50 mmHg (lit.)

mp

22-23 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(C)cc1OC

InChI

1S/C9H12O2/c1-7-4-5-8(10-2)9(6-7)11-3/h4-6H,1-3H3

InChI key

GYPMBQZAVBFUIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dimethoxytoluene is the main component of essential oil isolated from Phoenix dactylifera L. It was selectively oxidized to the corresponding 1,4-benzoquinones in the presence of hydrogen peroxide/methyltrioxorhenium in 1-butyl-3-methylimidazolium tetrafluoroborate.

Application

3,4-Dimethoxytoluene was used as an internal standard in HPLC quantifications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Convenient oxidation of alkylated phenols and methoxytoluenes to antifungal 1, 4-benzoquinones with hydrogen peroxide (H2O2)/methyltrioxorhenium (CH3 ReO3) catalytic system in neutral ionic liquid.
Bernini R, et al.
Tetrahedron, 62(33), 7733-7737 (2006)
Dimitris S Argyropoulos et al.
Journal of agricultural and food chemistry, 50(4), 658-666 (2002-02-07)
To define the abundance and comprehend the reactivity of dibenzodioxocins in lignin, model compound studies, specific degradation experiments on milled wood lignin, and molecular modeling calculations have been performed. Quantitative (31)P NMR measurements of the increase of biphenolic hydroxyl groups
Betül Demirci et al.
Acta tropica, 128(3), 557-560 (2013-08-21)
Date palm, Phoenix dactylifera L. (Arecaceae), grows commonly in the Arabian Peninsula and is traditionally used to treat various diseases. The aim of the present study was to identify chemical composition of the essential oil and to investigate the repellent
Evon Powell et al.
International journal of nanomedicine, 2(3), 449-459 (2007-11-21)
The interaction of the important but often overdosed local anesthetic bupivacaine, its structural analogs 2,6-dimethylaniline, and N-methyl-2,6-dimethylacetanilide, and cocaine, with several electron deficient aromatic moieties were studied primarily by proton NMR and UV-visible spectroscopy. In solution, the anesthetic, its analogs
Ting Zhang et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-11)
The unpleasant stale note is a negative factor hindering the consumption of instant ripened Pu-erh tea products. This study focused on investigating volatile chemicals in instant ripened Pu-erh tea that could mask the stale note via sensory evaluation, gas chromatography-mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service