Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

162434

Sigma-Aldrich

4-Chloro-2-phenylquinazoline

97%

Synonym(s):

AM-ex-OL, NSC 400965

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClN2
CAS Number:
Molecular Weight:
240.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

mp

124-126 °C (lit.)

SMILES string

Clc1nc(nc2ccccc12)-c3ccccc3

InChI

1S/C14H9ClN2/c15-13-11-8-4-5-9-12(11)16-14(17-13)10-6-2-1-3-7-10/h1-9H

InChI key

OBHKONRNYCDRKM-UHFFFAOYSA-N

Application

4-Chloro-2-phenylquinazoline was used in synthesis of axially chiral quinazoline-containing phosphinamine ligand, N,N,2-triphenylquinazolin-4-amine, sulfanyl and sulfinylbenzodiazines. It was used as reagent for the conversion of phenols to anilines.
Reactant involved in the synthesis of biologically active molecules including:
  • Nitrotriazole amines or nitroimidazole amines for use as antitrypanosomal activity and mammalian cytotoxicity
  • Quinazoline-containing piperazinylpyrimidine derivatives with antitumor activity
  • Quinazoline substituted cyclopentane as HCV NS3/4A protease inhibitors
  • Quinazolines with antibacterial and antitumor activity
  • Aurora inhibitor MK-0457

Reactant involved in Suzuki-Miyaura cross-coupling and catalyst-free / base-free water promoted nucleophilic aromatic substitution

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 37, 1681-1681 (1972)
The Journal of Organic Chemistry, 29, 1893-1893 (1964)
A highly efficient red electrophosphorescent iridium (iii) complex containing phenyl quinazoline ligand in polymer light-emitting diodes.
Mei Q, et al.
Journal of Materials Chemistry, 22(14), 6878-6884 (2012)
Syntheses of sulfoxide derivatives in the benzodiazine series. Diazines. Part 37.
Le Fur N, et al.
Tetrahedron, 60(36), 7983-7994 (2004)
The preparation and resolution of 2-phenyl-Quinazolinap, a new atropisomeric phosphinamine ligand for asymmetric catalysis.
McCarthy M, et al.
Tetrahedron Asymmetry, 10(14), 2797-2807 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service