Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161985

Sigma-Aldrich

p-Chloromercuriobenzoate

Synonym(s):

4-(Hydroxymercurio)benzoic acid sodium salt, p-Hydroxymercuribenzoic acid sodium salt, PCMB, Sodium 4-(hydroxymercurio)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-HOHgC6H4CO2Na
CAS Number:
Molecular Weight:
360.69
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

[Na+].O[Hg]c1ccc(cc1)C([O-])=O

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Campanella et al.
Analytical biochemistry, 483, 27-33 (2015-05-07)
The aim of our study was to investigate how denaturing agents commonly used in protein analysis influence the labeling between a reactive molecule and proteins. For this reason, we investigated the labeling of ovalbumin (OVA) as a globular model protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service