Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

160369

Sigma-Aldrich

Phenylmalonic acid

98%

Synonym(s):

α-Carboxyphenylacetic acid, α-Phenylmalonic acid, 2-Phenylpropanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(COOH)2
CAS Number:
Molecular Weight:
180.16
Beilstein:
2048729
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

mp

149-152 °C (dec.) (lit.)

SMILES string

OC(=O)C(C(O)=O)c1ccccc1

InChI

1S/C9H8O4/c10-8(11)7(9(12)13)6-4-2-1-3-5-6/h1-5,7H,(H,10,11)(H,12,13)

InChI key

WWYDYZMNFQIYPT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V L Lapidus et al.
Antibiotiki, 26(5), 357-360 (1981-05-01)
Distribution of carbenicillin, benzylpenicillin and phenylmalonic acid in butyl acetate-water and methylene chloride-water systems within wide ranges of pH of the aqueous phase was studied. The experimental data were reviewed taking into account the distribution between the phases of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service