Skip to Content
MilliporeSigma
All Photos(1)

Documents

159018

Sigma-Aldrich

β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

98%

Synonym(s):

1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H24O9
CAS Number:
Molecular Weight:
504.48
Beilstein:
100243
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +24.3°, c = 1 in pyridine

mp

128-130 °C (lit.)

SMILES string

CC(=O)O[C@@H]1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@H]1OC(=O)c4ccccc4

InChI

1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28-/m1/s1

InChI key

GCZABPLTDYVJMP-CBUXHAPBSA-N

Application

Starting material for nucleoside synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service