Skip to Content
MilliporeSigma
All Photos(1)

Documents

157937

Sigma-Aldrich

Dimethyl 2-oxoheptylphosphonate

technical grade, 85%

Synonym(s):

(2-Oxoheptyl)phosphonic acid dimethyl ester, Dimethyl (hexanoylmethyl)phosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COCH2P(O)(OCH3)2
CAS Number:
Molecular Weight:
222.22
Beilstein:
1946305
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.444 (lit.)

bp

109 °C/0.4 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(=O)CP(=O)(OC)OC

InChI

1S/C9H19O4P/c1-4-5-6-7-9(10)8-14(11,12-2)13-3/h4-8H2,1-3H3

InChI key

LQZCYXCHWNQBKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Beirut reaction for the preparation of phosphonylated quinoxaline dioxides
  • Preparation of pyrrolomorphinan derivatives as κ opioid agonists via cyclocondensation, Vilsmeier-Haack formylation, Horner-Wadsworth-Emmons reaction, and Kocienski-modified Julia olefination from naltrexone methyl ether
  • Synthesis of C-glycosides amphiphiles via solvent-free Horner-Wadsworth-Emmons reaction with unprotected sugars
  • Preparation of specific inhibitors of endocannabinoid biosynthesis
A key intermediate in prostaglandin synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service