Skip to Content
MilliporeSigma
All Photos(1)

Documents

156558

Sigma-Aldrich

α-Methoxy-α-(trifluoromethyl)phenylacetic acid

97%

Synonym(s):

MTPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OCH3)(CF3)CO2H
CAS Number:
Molecular Weight:
234.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.476 (lit.)

bp

95-98 °C/0.05 mmHg (lit.)

mp

41-46 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(C(O)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H9F3O3/c1-16-9(8(14)15,10(11,12)13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)

InChI key

JJYKJUXBWFATTE-UHFFFAOYSA-N

Application

Diastereomeric bis-esters of (±)-trans-7,8-dihydroxy-7,8,9,10-tetrahydrobenzo[α]pyrene with MTPA has been used as a noncarcinogenic precursor to obtain the (+)and (-)-dihydrodiols by chromatographic resolution. It has also been used to develop empirically derived correlation of configuration and nmr chemical shifts for diastereomeric MTPA esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear Magnetic Resonance Enantiomer Reagents. Configurational Correlations via Nuclear Magnetic Resonance Chemical Shifts of Diastereomeric Mandelate, 0-Methylmandelate, and α-Methoxy-α-trifluoromethylphenylacetate (MTPA) Esters
Dale JA and Mosher HS
Journal of the American Chemical Society, 95(2), 512-519 (1973)
Yuka Uemura et al.
Chemical & pharmaceutical bulletin, 63(8), 608-616 (2015-08-04)
From the leaves of Meliosma lepidota ssp. squamulata, megastigmane glucosides with spiro-structures and megastigmanes were isolated. Their structures were determined by X-ray crystallographic analyses and spectroscopic investigation. The absolute structures of the megastigmanes were determined by the modified Mosher's method.
W Levin et al.
The Journal of biological chemistry, 255(19), 9067-9074 (1980-10-10)
The (+)- and (-)-enantiomers of benzo[a]pyrene 7,8-oxide are hydrated stereospecifically at C-8 to (-)- and (+)-trans-7,8-dihydroxy-7,8-dihydrobenzo[a]pyrene, respectively, by rat hepatic epoxide hydrolase. The (-)-enantiomer of benzo[a]pyrene 7,8-oxide is metabolized by microsomal epoxide hydrolase at a rate 3- to 4-fold greater
Dongsheng Du et al.
Fitoterapia, 101, 73-79 (2015-01-07)
Two new germacrane sesquiterpenes, yedoensins A (1) and B (2), together with 8 known ones (3-10) were isolated from the herb of Viola yedoensis. The structures of the new compounds were established by extensive spectroscopic means including 1D ((1)H and
Guangyu Wang et al.
Oncology letters, 10(5), 2835-2841 (2016-01-02)
MicroRNA-181 (miR-181) has been recently demonstrated to participate in the differentiation and development of immune cells, including natural killer cells and B and T lymphocytes, and myeloid linages, including erythroid and megakaryocytic cells. The aberrant expression of miR-181, particularly low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service