Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

156493

Sigma-Aldrich

2-Acetyl-1,3-cyclohexanedione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H7(=O)2
CAS Number:
Molecular Weight:
154.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

85 °C/0.1 mmHg (lit.)

mp

20 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CC(=O)C1C(=O)CCCC1=O

InChI

1S/C8H10O3/c1-5(9)8-6(10)3-2-4-7(8)11/h8H,2-4H2,1H3

InChI key

CHNXDYRMRBQOEF-UHFFFAOYSA-N

Application

The product has been used in the chelation of aqueous iron(III) that is present in alcoholic drinks, such as wine and beer. It affects beer fermentation, stability and quality. It has also been used to evaluate mesotrione assay specificity as its structure is similar to mesotrione (a triketone herbicide).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chelation of aqueous iron(III) by 2-acetyl-1,3-cyclohexanedione and beer ageing
Blanco CA, et al.
Food Chemistry, 81(4), 561-568 (2003)
Christos Mastichiadis et al.
The Analyst, 128(4), 404-410 (2003-05-14)
A straightforward, low-cost fluoroimmunoassay (FIA) for the determination of the new triketone herbicide mesotrione has been developed and optimized. The protein-mesotrione conjugate, immobilized on white opaque microtitration wells competes with the mesotrione in the sample or standard for the limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service