Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

156140

Sigma-Aldrich

o-Phenylene phosphorochloridate

technical grade

Synonym(s):

2-Chloro-1,3,2-benzodioxaphosphole 2-oxide, o-Phenylene chlorophosphate, Catechol cyclophosphorochloridate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClO3P
CAS Number:
Molecular Weight:
190.52
Beilstein:
142368
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

bp

120 °C/9 mmHg (lit.)

SMILES string

ClP1(=O)Oc2ccccc2O1

InChI

1S/C6H4ClO3P/c7-11(8)9-5-3-1-2-4-6(5)10-11/h1-4H

InChI key

KMWSGKPLIWNTEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Uridine diphosphate-glucose derivatives as potential chain terminators of β-glucan biosynthesis with antifungal activity
  • Vinyloxy phosphorus monomers

  • Phosphorylation agent

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service