Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15514

Sigma-Aldrich

Boc-Trp-N-carboxyanhydride

≥98.0% (CHN)

Synonym(s):

Nα-Boc-L-tryptophan Nα-carboxy anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O5
CAS Number:
Molecular Weight:
330.34
Beilstein:
7082757
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98.0% (CHN)

optical activity

[α]20/D +78±5°, c = 1% in THF

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

~135 °C (dec.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1C(Cc2c[nH]c3ccccc23)C(=O)OC1=O

InChI

1S/C17H18N2O5/c1-17(2,3)24-16(22)19-13(14(20)23-15(19)21)8-10-9-18-12-7-5-4-6-11(10)12/h4-7,9,13,18H,8H2,1-3H3/t13-/m0/s1

InChI key

WSFZLNNHQNYZNF-ZDUSSCGKSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service