Skip to Content
MilliporeSigma
All Photos(1)

Documents

15505

Sigma-Aldrich

Boc-Thr-OH

≥98.0% (T)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-threonine, Boc-L-threonine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
219.23
Beilstein:
2051735
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98.0% (T)

optical activity

[α]20/D −8.5±1°, c = 1% in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

80-82 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C9H17NO5/c1-5(11)6(7(12)13)10-8(14)15-9(2,3)4/h5-6,11H,1-4H3,(H,10,14)(H,12,13)/t5-,6+/m1/s1

InChI key

LLHOYOCAAURYRL-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service