Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154040

Sigma-Aldrich

4-Fluoro-α-methylbenzylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH(CH3)NH2
CAS Number:
Molecular Weight:
139.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 154040, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

96%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

145 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

CC(N)c1ccc(F)cc1

InChI

1S/C8H10FN/c1-6(10)7-2-4-8(9)5-3-7/h2-6H,10H2,1H3

InChI key

QGCLEUGNYRXBMZ-UHFFFAOYSA-N

Application

4-Fluoro-α-methylbenzylamine was used in the synthesis of novel thioureas with diuretic and saluretic activity.[1] It has also been used for nucleophilic substitution reactions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eur. J. Med. Chem. - Chem. Ther., 15, 41-41 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service