153788
Dodecanoic acid
≥99.5%
Synonym(s):
ABL, Lauric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
Beilstein:
1099477
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
vapor pressure
1 mmHg ( 121 °C)
50 mmHg ( 210 °C)
Assay
≥99.5%
bp
225 °C/100 mmHg (lit.)
mp
44-46 °C (lit.)
density
0.883 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCC(O)=O
InChI
1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)
InChI key
POULHZVOKOAJMA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Isaac Levy et al.
The Journal of clinical endocrinology and metabolism, 100(11), 4106-4113 (2015-09-01)
Adults with hypoparathyroidism have significant rates of nephrocalcinosis and impaired renal function. Little is known about the impact of hypoparathyroidism treatment on renal function in children. To determine the prevalence and predictors for renal abnormalities (nephrocalcinosis and decreased estimated glomerular
Esther Mesman et al.
Bipolar disorders, 17(1), 39-49 (2014-07-22)
There is increasing evidence that both immune and neurochemical alterations are involved in the pathogenesis of bipolar disorder; however, their precise role remains unclear. In this study, we aimed to evaluate neuro-immune changes in a prospective study on children of
Juliette Lambert et al.
Oncotarget, 5(15), 6280-6288 (2014-07-16)
We analysed the prognostic significance of minimal residual disease (MRD) level in adult patients with acute myeloid leukemia (AML) treated in the randomized gemtuzumab ozogamicin (GO) ALFA-0701 trial. Levels of WT1 and NPM1 gene transcripts were assessed using cDNA-based real-time
Meng-Meng Bai et al.
Journal of agricultural and food chemistry, 63(8), 2198-2205 (2015-02-14)
Eucommia ulmoides leaves have been used as a functional food and drink in China. The purpose of this study was to identify the bioactive constituents with soluble epoxide hydrolase (sEH) inhibitory activity and anti-inflammatory properties. Twenty-seven known compounds (1-27) were
M Schmidt et al.
Leukemia, 28(12), 2292-2299 (2014-09-13)
To study clonal evolution in chronic myeloid leukemia (CML), we searched for BCR-ABL-independent gene mutations in both Philadelphia chromosome (Ph)-negative and Ph-positive clones in 29 chronic-phase CML patients by targeted deep sequencing of 25 genes frequently mutated in myeloid disorders.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service