Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

153117

Sigma-Aldrich

1-Benzyl-3-piperidone hydrochloride hydrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO · HCl · xH2O
CAS Number:
Molecular Weight:
225.71 (anhydrous basis)
Beilstein:
4334779
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

functional group

ketone
phenyl

SMILES string

Cl[H].[H]O[H].O=C1CCCN(C1)Cc2ccccc2

InChI

1S/C12H15NO.ClH.H2O/c14-12-7-4-8-13(10-12)9-11-5-2-1-3-6-11;;/h1-3,5-6H,4,7-10H2;1H;1H2

InChI key

XDAGZUGDGORLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Benzyl-3-piperidone hydrochloride hydrate was used to prepare 7-Benzyl-3-(phenylthio)-1-oxa-4,7-diazaspiro[4.5]dec-3-en-2-one[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Liang et al.
Tetrahedron letters, 50(49), 6810-6813 (2010-02-18)
Microwave irradiation facilitated the synthesis of 4-arylthio-3-oxazolin-5-ones from ethyl cyanoformate, thiophenol, and cyclic ketones. Subsequent decarboxylation and in situ [3+2] cycloaddition provided novel 2,3,4,5-tetrahydro-1H-pyrrolo[1,2-c][1,3]diazepine scaffolds after a spontaneous retro-Mannich domino reaction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service