Skip to Content
MilliporeSigma
All Photos(1)

Documents

152722

Sigma-Aldrich

5,6,11,12-Tetrahydrodibenz[b,f]azocin-6-one

97%

Synonym(s):

11,12-Dihydrodibenz[b,f]azocin-6(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

246-248 °C (lit.)

SMILES string

O=C1Nc2ccccc2CCc3ccccc13

InChI

1S/C15H13NO/c17-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)16-15/h1-8H,9-10H2,(H,16,17)

InChI key

JZDBKFVASLJURR-UHFFFAOYSA-N

Application

5,6,11,12-Tetrahydrodibenz[b,f]azocin-6-one (11,12-Dihydrodibenz[b,f]azocin-6(5H)-one) was used in the preparation of 5-Benzyl-11,12-dihydrodibenz[b,f]azocin-6(5H)-one.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational behaviour of medium-sized rings. Part I. 5, 6, 11, 12-Tetrahydrodibenzo [a, e] cyclo-octene (1, 2, 5, 6-dibenzocyclo-octa-1, 5-diene) and heterocyclic analogues.
Crossley R, et al.
Journal of the Chemical Society. Perkin Transactions 1, 205-217 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service