Skip to Content
MilliporeSigma
All Photos(1)

Documents

152153

Sigma-Aldrich

2-Formylphenoxyacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H4OCH2CO2H
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

129.5-132 °C (lit.)

SMILES string

OC(=O)COc1ccccc1C=O

InChI

1S/C9H8O4/c10-5-7-3-1-2-4-8(7)13-6-9(11)12/h1-5H,6H2,(H,11,12)

InChI key

ANWMNLAAFDCKMT-UHFFFAOYSA-N

Application

2-Formylphenoxyacetic acid was used to synthesize azomethine derivatives. It was also used in the preparation of ligands L(1)H(2) and L(2)H(2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priya P Netalkar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 762-770 (2012-08-21)
A series of novel Co(II), Ni(II), Cu(II) and Zn(II) complexes of tri and tetratopic hydrazones have been prepared. Ligands L(1)H(2) and L(2)H(2) were synthesized by the condensation of 2-formylphenoxyacetic acid with 2-hydrazinobenzothiazole and 2-hydroxy-3-hydrazinebenzopyrazine, respectively. The prepared complexes were characterized
Amjid Iqbal et al.
Molecules (Basel, Switzerland), 12(2), 245-254 (2007-09-12)
A series of eight new azomethine derivatives were synthesized by reacting 2-formylphenoxyacetic acid with aromatic amines. The chemical structures of these compounds were confirmed by means of 1H-NMR, 13C-NMR, MS and elemental analysis. The compounds were assayed by the disc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service