Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

151300

Sigma-Aldrich

Flufenamic acid

97%

Synonym(s):

N-(3-[Trifluoromethyl]phenyl)anthranilic acid, N-(α,α,α-Trifluoro-m-tolyl)anthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CF3C6H4NH)C6H4CO2H
CAS Number:
Molecular Weight:
281.23
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

mp

132-135 °C (lit.)

SMILES string

OC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F

InChI

1S/C14H10F3NO2/c15-14(16,17)9-4-3-5-10(8-9)18-12-7-2-1-6-11(12)13(19)20/h1-8,18H,(H,19,20)

InChI key

LPEPZBJOKDYZAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Klang et al.
Pharmaceutics, 3(2), 275-306 (2011-01-01)
Mild non-ionic sucrose ester surfactants can be employed to produce lipid-based drug delivery systems for dermal application. Moreover, sucrose esters of intermediate lipophilicity such as sucrose stearate S-970 possess a peculiar rheological behavior which can be employed to create highly
Fei Ding et al.
Physical chemistry chemical physics : PCCP, 18(9), 6595-6606 (2016-02-13)
Ligand-protein recognition is increasingly explored through different biophysical approaches, but the effects of protein flexibility are frequently neglected in many applications. The main goal of this contribution is to comparatively study the flexibility and dynamics of homologous model proteins, i.e.
Eleanor R Turpin et al.
Molecular pharmaceutics, 15(10), 4654-4667 (2018-08-25)
The problem of predicting small molecule-polymer compatibility is relevant to many areas of chemistry and pharmaceutical science but particularly drug delivery. Computational methods based on Hildebrand and Hansen solubility parameters, and the estimation of the Flory-Huggins parameter, χ, have proliferated
Weifan Jiang et al.
International immunopharmacology, 86, 106723-106723 (2020-07-03)
Diclofenac (DCF) is a widely used nonsteroidal anti-inflammatory drug, but it comes with a high risk of drug-induced liver injury (DILI). Despite the quinone-imine adduct pathways, the immunotoxicity is recently considered as another factor for DILI. However, such immune responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service