Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150738

Sigma-Aldrich

Azobenzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H5
CAS Number:
Molecular Weight:
182.22
Beilstein:
1819138
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

vapor pressure

1 mmHg ( 104 °C)

Assay

96%

autoignition temp.

890 °F

bp

293 °C (lit.)

mp

65-68 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

c1ccc(cc1)\N=N\c2ccccc2

InChI

1S/C12H10N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H/b14-13+

InChI key

DMLAVOWQYNRWNQ-BUHFOSPRSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fastest thermal isomerization of an azobenzene for nanosecond photoswitching applications under physiological conditions.
Jaume Garcia-Amorós et al.
Angewandte Chemie (International ed. in English), 51(51), 12820-12823 (2012-11-13)
P S Ramanujam et al.
Optics express, 21(2), 1812-1819 (2013-02-08)
We demonstrate a new type of anisotropy in thin films of amorphous azobenzene polymers induced between 570 and 633 nm, where the absorbance in the film is on the order of 0.05. The anisotropy has a pronounced radial contribution. This
Laura Maggini et al.
Nanoscale, 5(2), 634-645 (2012-12-12)
Photothermally responsive supramolecular polymers containing azobenzene units have been synthesised and employed as dispersants for multi-walled carbon nanotubes (MWCNTs) in organic solvents. Upon triggering the trans-cis isomerisation of the supramolecular polymer intermolecular interactions between MWCNTs and the polymer are established
C Renner et al.
The journal of peptide research : official journal of the American Peptide Society, 65(1), 4-14 (2005-02-03)
Over the last decades azobenzene has been the most widely used optical trigger for the synthesis of photoresponsive systems ranging from poly-alpha-amino acids to innovative materials with light-controlled mechanical and optical properties. More recently, its use in form of appropriate
Daisuke Ishikawa et al.
Langmuir : the ACS journal of surfaces and colloids, 29(14), 4622-4631 (2012-12-20)
The growth processes of self-assembled monolayers (SAMs) of two azobenzene disulfides formed on flat gold surfaces were studied to confirm the effect of the intermolecular interactions between azobenzene molecules on the light-triggered surface morphologies of the SAMs. Scanning tunneling microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service